Skip to Content
Merck

Skip To

74382

N-Heptafluorobutyrylimidazole

derivatization grade (GC derivatization), LiChropur, ≥98.0% (GC)

Synonym(s):

1-(Perfluorobutyryl)imidazole, NSC 151966

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
CZK 2,050.00
10 mL
Please contact Customer Service for Availability
CZK 14,200.00
10 x 1 mL
Please contact Customer Service for Availability
CZK 16,900.00

About This Item

Empirical Formula (Hill Notation):
C7H3F7N2O
CAS Number:
Molecular Weight:
264.10
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
23151817
EC Number:
251-063-8
MDL number:
Assay:
≥98.0% (GC), 98.5-101.0% (wt., NT)
Form:
liquid

CZK 2,050.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

derivatization grade (GC derivatization)

Quality Segment

assay

≥98.0% (GC), 98.5-101.0% (wt., NT)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.3865 (lit.)

bp

161 °C (lit.)

density

1.490 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(=O)n1ccnc1

InChI

1S/C7H3F7N2O/c8-5(9,6(10,11)7(12,13)14)4(17)16-2-1-15-3-16/h1-3H

InChI key

MSYHGYDAVLDKCE-UHFFFAOYSA-N

Application

N-Heptafluorobutyrylimidazole may be used as a derivatizing agent in the analysis of aminoglycoside drugs such as gentamicin, tobramycin, netihnicin, and amikacin in samples of human serum using gas chromatography (GC).[1]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
902577987943504
assay

≥98.0% (GC), 98.5-101.0% (wt., NT)

assay

≥98.5% (GC)

assay

≥98.5% (GC)

assay

99.0-101.0% (wt., AT)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

form

liquid

form

liquid

form

liquid

form

-

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-

refractive index

n20/D 1.3865 (lit.)

refractive index

n20/D 1.471 (lit.)

refractive index

n20/D 1.288 (lit.)

refractive index

-


Still not finding the right product?

Explore all of our products under N-Heptafluorobutyrylimidazole


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters