Skip to Content
Merck
All Photos(1)

Key Documents

73091

Sigma-Aldrich

Potassium sulfate solution

BioUltra, 0.5 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
MDL number:
UNSPSC Code:
12352200

product line

BioUltra

concentration

0.5 M in H2O

impurities

insoluble matter, passes filter test
≤0.0005% total nitrogen (N)

pH

5.5-7.5 (20 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

density

1.056 g/mL at 20 °C

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤20 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤20 mg/kg
Zn: ≤1 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.01
λ: 280 nm Amax: ≤0.01

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Scarano et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 68(5), 964-968 (2010-03-13)
Calcium sulfate (CaS) is a simple, biocompatible material with a very long, safe clinical history in several different fields of medicine. CaS is a rapidly resorbing material that leaves behind a calcium phosphate lattice, which promotes bone regeneration and hemostasis.
Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
Ashley C Parker et al.
Clinical orthopaedics and related research, 469(11), 3008-3015 (2011-05-10)
Local drug delivery has substantial potential to prevent infections compared with systemic delivery. Although calcium sulfate (CaSO(4)) has been studied for local drug delivery and two types are commercially available, it is unknown whether they differentially release antibiotics. We determined
Jingjing Zheng et al.
Bioresource technology, 110, 224-231 (2012-02-15)
This study investigated the preparation and characteristics of protein-coated microcrystals (PCMCs) from Pseudomonas cepacia lipase (PS) and K(2)SO(4), and their application in biodiesel synthesis, via single factorial experiments and response surface methodology (RSM), the optimized PCMC-PS exhibited high activity and
C C Constantinoiu et al.
Poultry science, 86(9), 1910-1914 (2007-08-21)
Immunoglobulin Y is different from most of the other immunoglobulins because it does not bind protein A or protein G. Thiophilic gel chromatography has been successfully used to purify IgY from chicken egg yolk, but the technology has not previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service