Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.74
UNSPSC Code:
41171614
form
powder
Quality Segment
shelf life
limited shelf life, expiry date on the label
application(s)
microbiology
storage temp.
2-8°C
Application
An antibiotic supplement recommended for selective isolation of Pseudomonas aeruginosa.
Preparation Note
Rehydrate the contents of 1 vial aseptically with 5 ml of sterile distilled water. Mix well and aseptically add it to 1000 ml of sterile, molten cooled (45-50ºC) M-PA Agar Base (Cat. No.42800). Mix well and aseptically pour into sterile petri plates.
Other Notes
Composition
(per vial sufficient for 1000 mL medium)
Sulphapyridine 176mg
Kanamycin 8.5mg
Nalidixic acid 37mg
Amphotericin B 5mg
(per vial sufficient for 1000 mL medium)
Sulphapyridine 176mg
Kanamycin 8.5mg
Nalidixic acid 37mg
Amphotericin B 5mg
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.