71013BC
GDS Top 6 STEC (eae) Tq
BioControl, Molecular based PCR test for the detection of the eae gene in "Top Six" non-O157 Shiga Toxigenic E. coli (STEC) in food
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
shelf life
6‑12 mo.
manufacturer/tradename
BioControl Systems 71013-100
application(s)
food and beverages
compatibility
BioControl GDS
storage temp.
2-8°C
General description
GDS Top 6 STEC Tq is an automated nucleic acid amplification system for the detection of the eae gene in the following select O-serogroups of E. coli: O103, O111, O121, O145, O26 and O45. Assurance GDS utilizes a proprietary IMS-based sample preparation procedure to isolate organisms belonging to these specific 6 O-serogroups prior to genetic analysis for the presence of the eae gene.
Features and Benefits
- Sequential approach to detecting Top STEC. Following sample preparation with IMS against the Top 6 O groups, samples are analyzed for the presence of (eae) with the GDS Top 6 STEC (eae) assay. Positive samples are subsequently analyzed for Shiga Toxin genes with the GDS Shiga Toxin Genes (Top 6) assay.
- Single-step 10-18 h enrichment in mEHEC, enrichment media common to all BioControl methods for E.coli O157:H7, Shiga Toxin genes, and Top STEC
Components
- Top STEC (eae) Amplification Tubes Tq
- Top 6 STEC Concentration Reagent
- Resuspension Buffer Tq
- Top STEC Wash Solution
Compatibility
BioControl GDS
Testing Method
PCR, Immunomagnetic Separation (IMS)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service