Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mL | Please contact Customer Service for Availability | CZK 5,760.00 |
About This Item
product line
Selectophore™
Quality Segment
form
liquid
composition
Calcium ionophore IV, 0.072 wt. % (21198), 2-Nitrophenyl octyl ether, 4.748 wt. % (73732), Poly(vinyl chloride), 2.379 wt. % (81392), Potassium tetrakis(4-chlorophenyl)borate, 0.022 wt. % (60591), Tetrahydrofuran, 92.780 wt. % (87369)
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form solid |
| composition Calcium ionophore IV, 0.072 wt. % (21198), Poly(vinyl chloride), 2.379 wt. % (81392), Tetrahydrofuran, 92.780 wt. % (87369), 2-Nitrophenyl octyl ether, 4.748 wt. % (73732) | composition Calcium ionophore I, 10.0 wt. % (21192), 2-Nitrophenyl octyl ether, 89.0 wt. % (73732), Sodium tetraphenylborate, 1.0 wt. % (72018) | composition Hydrogen ionophore I, 10.0 wt. % (95292), 2-Nitrophenyl octyl ether, 89.3 wt. % (73732), Potassium tetrakis(4-chlorophenyl)borate, 0.7 wt. % (60591) | composition - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| product line Selectophore™ | product line Selectophore™ | product line Selectophore™ | product line Selectophore™ |
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Respiratory system
supp_hazards
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
-0.4 °F - closed cup
flash_point_c
-18.0 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






