Skip to Content
Merck

70165

Liver Powder, dehydrated

BioChemika, suitable for microbiology

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioChemika

shelf life

limited shelf life, expiry date on the label

pH

~6 (2% in H2O)

application(s)

microbiology

Preparation Note

To prepare a liver infusion medium, add 50 g of liver powder to 1 litre of distilled water and allow to infuse (with frequent agitation) for 1 hour at 50°C. Boil the mixture for a few minutes to coagulate protein, strain through 60 mesh stainless steel gauze, add 10 g of peptone and 5 g of sodium chloride. Adjust the reaction to pH 7.2, boil, strain through gauze as above, and sterilize by autoclaving at 121°C for 15 minutes.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library