Skip to Content
Merck
All Photos(1)

Documents

70162

Sigma-Aldrich

Nalidixic acid

≥99.0% (T)

Synonym(s):

1,4-Dihydro-1-ethyl-7-methyl-1,8-naphthyridin-4-one-3-carboxylic acid, 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
Beilstein:
750515
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥99.0% (T)

mp

227-229 °C (lit.)
228-230 °C

solubility

chloroform: 20 mg/mL, clear

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

CCN1C=C(C(O)=O)C(=O)c2ccc(C)nc12

InChI

1S/C12H12N2O3/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14/h4-6H,3H2,1-2H3,(H,16,17)

InChI key

MHWLWQUZZRMNGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Other Notes

Inhibitor of bacterial DNA synthesis

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the mode of action of nalidixic acid.
A M Pedrini et al.
European journal of biochemistry, 25(2), 359-365 (1972-02-15)
Christopher M Parry
Transactions of the Royal Society of Tropical Medicine and Hygiene, 98(7), 413-422 (2004-05-13)
Multidrug-resistant (MDR) Salmonella Typhi (resistant to chloramphenicol, ampicillin, and trimethoprim-sulphamethoxazole) and isolates with reduced susceptibility to fluoroquinolones (indicated by resistance to nalidixic acid, NaR) have caused epidemics and become endemic in southern Viet Nam during the 1990s. Short courses of
Robert J Commons et al.
The Medical journal of Australia, 196(5), 332-336 (2012-03-22)
To determine incidence and trends in antibiotic resistance in Australian Salmonella enterica subspecies enterica serovars Typhi (S. Typhi) and Paratyphi (S. Paratyphi) isolates over the past 26 years. A retrospective analysis of consecutive microbiologically confirmed enteric fever isolates. All S.
Migla Miskinyte et al.
Antimicrobial agents and chemotherapy, 57(1), 189-195 (2012-10-24)
Mutations causing antibiotic resistance usually incur a fitness cost in the absence of antibiotics. The magnitude of such costs is known to vary with the environment. Little is known about the fitness effects of antibiotic resistance mutations when bacteria confront
George X Song-Zhao et al.
Mucosal immunology, 7(4), 763-774 (2013-11-28)
Polymorphisms in the intracellular pattern recognition receptor gene NLRP3 (NLR family, pyrin domain containing 3) have been associated with susceptibility to Crohn's disease, a type of inflammatory bowel disease. Following tissue damage or infection, NLRP3 triggers the formation of inflammasomes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service