Skip to Content
Merck
All Photos(1)

Documents

70112

Supelco

AQUANAL-professional Vario H Chlorine Free (DPD)

determination of chlorine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

form

powder

measuring range

0.01-2 mg/L (Cl2)

application(s)

environmental
general analytical

Packaging

100 powder packs for 10 mL sample volume

Analysis Note

method: N,N-Diethyl-p-phenylenediamine (DPD)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service