Skip to Content
Merck
All Photos(1)

Key Documents

70061

Supelco

AQUANAL-professional DPD No. 1

determination of bromine, chlorine, chlorine dioxide and ozone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

form

tablet

application(s)

environmental
general analytical

Packaging

100 tablets

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service