Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H14.5NO4S · 0.5Na
CAS Number:
Molecular Weight:
220.25
UNSPSC Code:
12161700
assay
≥97.0% (T)
impurities
≤3% water
useful pH range
6.5-7.9
pKa (25 °C)
7.2
solubility
H2O: 0.5 g/mL, clear, colorless
SMILES string
[Na+].OS(=O)(=O)CCCN1CCOCC1.[O-]S(=O)(=O)CCCN2CCOCC2
InChI
1S/2C7H15NO4S.Na/c2*9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h2*1-7H2,(H,9,10,11);/q;;+1/p-1
InChI key
OVCWZKQPQOVIDT-UHFFFAOYSA-M
Still not finding the right product?
Explore all of our products under MOPS hemisodium salt
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.