Skip to Content
Merck

Skip To

68706

Perfluorotridecanoic acid

analytical standard

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
F3C(CF2)11CO2H
CAS Number:
Molecular Weight:
664.11
UNSPSC Code:
41116107
MDL number:


grade

analytical standard

assay

≥98.0% (GC)

form

solid

shelf life

limited shelf life, expiry date on the label

mp

112-123 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C13HF25O2/c14-2(15,1(39)40)3(16,17)4(18,19)5(20,21)6(22,23)7(24,25)8(26,27)9(28,29)10(30,31)11(32,33)12(34,35)13(36,37)38/h(H,39,40)

InChI key

LVDGGZAZAYHXEY-UHFFFAOYSA-N

General description

This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)


Still not finding the right product?

Explore all of our products under Perfluorotridecanoic acid


signalword

Danger

target_organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1 - vPvB


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

72629-94-8

CAS No.

72629-94-8

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters