Skip to Content
Merck

68541

N,N′-Bis(salicylidene)-o-phenylenediamine Vanadium(IV) oxide Complex

≥98.0% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H14N2O3V
CAS Number:
Molecular Weight:
381.28
NACRES:
NB.61
PubChem Substance ID:
UNSPSC Code:
26111700
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

solid

reaction suitability

reagent type: catalyst
core: vanadium

storage temp.

2-8°C

SMILES string

O=[V]1Oc2ccccc2\C=N\c3ccccc3\N=C\c4ccccc4O1

InChI

1S/C20H16N2O2.O.V/c23-19-11-5-1-7-15(19)13-21-17-9-3-4-10-18(17)22-14-16-8-2-6-12-20(16)24;;/h1-14,23-24H;;/q;;+2/p-2/b21-13+,22-14+;;

InChI key

GZMVPBRTLSLWAO-FDOBHKDXSA-L

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Used in the direct determination of hydrogen phosphate and in a indicator electrode for the determination of hydrogen phosphate with Ba(NO3)2


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library