Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
10 mg | Please contact Customer Service for Availability | CZK 1,530.00 |
About This Item
Linear Formula:
HO2CCH2C[CH(CH3)2](OH)CO2H
CAS Number:
Molecular Weight:
176.17
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
grade
analytical standard
assay
≥97.5% (HPLC)
form
solid
shelf life
limited shelf life, expiry date on the label
mp
138-145 °C, 144-146 °C (lit.)
SMILES string
CC(C)C(O)(CC(O)=O)C(O)=O
InChI
1S/C7H12O5/c1-4(2)7(12,6(10)11)3-5(8)9/h4,12H,3H2,1-2H3,(H,8,9)(H,10,11)
InChI key
BITYXLXUCSKTJS-UHFFFAOYSA-N
Other Notes
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form solid | form - | form powder or crystals |
| grade analytical standard | grade - | grade - | grade - |
| assay ≥97.5% (HPLC) | assay 99% | assay 98% | assay 98% |
| shelf life limited shelf life, expiry date on the label | shelf life - | shelf life - | shelf life - |
| mp 138-145 °C | mp 76-80 °C (lit.) | mp 144-146 °C (lit.) | mp 78-80 °C (lit.) |
Still not finding the right product?
Explore all of our products under 2-Isopropylmalic acid
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


