Skip to Content
Merck
All Photos(1)

Key Documents

67403

Supelco

Cirsilineol

analytical standard

Synonym(s):

4′,5-Dihydroxy-3′,6,7-trimethoxyflavone, 6-Methoxyluteolin 3′,7-dimethyl ether, Anisomelin, Eupatrin, Fastigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
Beilstein:
1355108
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC and enzymatic)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COc1cc(ccc1O)C2=CC(=O)c3c(O)c(OC)c(OC)cc3O2

InChI

1S/C18H16O7/c1-22-13-6-9(4-5-10(13)19)12-7-11(20)16-14(25-12)8-15(23-2)18(24-3)17(16)21/h4-8,19,21H,1-3H3

InChI key

VKOSQMWSWLZQPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cirsilineol is a bioactive flavone[1] isolated from the herb of Artemisia vestita Wall., widely used to treat a variety of inflammatory conditions such as contact dermatitis, rheumatoid arthritis and sepsis. Cirsilineol exhibits anti-proliferative activity, which is used against different cancer cells by inducing apoptosis.[2]

Application

Cirsilineol may be used as an analytical reference standard for the quantification of the analyte in vervain samples[1] and extracts of Teucrium polium[3] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-nian He et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 30(10), 761-763 (2005-08-04)
To study the chemical constituents of Anoectochilus roxburghii. Silica gel and sephadex LH-20 column chromotography were used in the isolation from the ethanol extracts of the whole plant, the compounds were determined on the basis of various modern spectroscopic analysis
H J Heo et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 8(3), 194-201 (2001-10-26)
Amyloid beta protein (Abeta)-induced free radical-mediated neurotoxicity is a leading hypothesis as a cause of Alzheimer's disease (AD). Abeta increased free radical production and lipid peroxidation in PC12 nerve cells, leading to apoptosis and cell death. The effect of 4',5-dihydroxy-3',6,7-trimethoxyflavone
Cirsilineol inhibits proliferation of cancer cells by inducing apoptosis via mitochondrial pathway
Sheng X, et al.
The Journal of Pharmacy and Pharmacology, 60(11), 39-44 (2008)
HPLC and UV-spectrophotometry analysis of flavonoids in spray-dried and freez-dried extracts of Teucrium polium L.(Lamiaceae)
Stefkov G, et al.
Macedonian pharmaceutical bulletin, 47(11), 39-44 (2012)
[Flavones isolated from Thymus carnosus Boiss].
E Maruhenda et al.
Annales pharmaceutiques francaises, 45(6), 467-470 (1987-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service