66411
Methylcyclohexanone
mixture of isomers, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
≥98.0% (GC)
composition
3-methylcyclohexanone, ~70%
4-methylcyclohexanone, ~30%
refractive index
n20/D 1.446
bp
167-169 °C (lit.)
density
0.914 g/mL at 20 °C (lit.)
SMILES string
CC1CCC(=O)CC1.CC2CCCC(=O)C2.CC3CCCCC3=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service