Skip to Content
Merck
All Photos(1)

Key Documents

62303

Sigma-Aldrich

Lipase from Candida lipolytica

powder (fine), ≥5 U/g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

biological source

fungus (Candida lipolytica)

Quality Level

form

fibers
powder (fine)

specific activity

≥5 U/g

mol wt

Mr ~26000

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyses 1 μmol of p-nitrophenyl butyrate per minute at 25 °C in 50 mM Tris-HCl buffer, at pH7.5, 10% acetonitrile.

Other Notes

For the biomodification of fats and oils[1]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Bati et al.
Journal of the American Oil Chemists' Society, 61, 1743-1743 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service