Skip to Content
Merck
All Photos(1)

Key Documents

62103

Supelco

Lewatit® TP 207

sodium form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

Quality Level

description

weakly acidic cation exchanger

form

particles (spherical)

moisture

53-58%

technique(s)

LPLC: suitable

loss

45-60% loss on drying, 110 °C

matrix

styrene-divinylbenzene (macroporous)

matrix active group

iminodiacetic acid functional group

particle size

400-1250 μm

capacity

2.2 meq/mL by wetted bed volume

separation technique

affinity

Application

Chelating resin especially suited for the selective binding of heavy metal cations from weakly acidic to weakly basic solutions.

Legal Information

Lewatit is a registered trademark of Lanxess Deutschland GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service