Skip to Content
Merck
All Photos(1)

Documents

62047

Supelco

Furfuryl methyl ether

analytical standard

Synonym(s):

2-(Methoxymethyl)furan, 2-Furylmethyl methyl ether, Methyl furfuryl ether, NSC 35554

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
Beilstein:
383598
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

storage temp.

2-8°C

InChI

1S/C6H8O2/c1-7-5-6-3-2-4-8-6/h2-4H,5H2,1H3

InChI key

GANSPRKOWQQXPE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service