Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
228-408-6
Beilstein/REAXYS Number:
2453103
MDL number:
grade
analytical standard
Quality Segment
assay
≥99.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
impurities
≤0.3% water
refractive index
n20/D 1.504-1.506, n20/D 1.505 (lit.)
bp
290 °C (lit.)
density
1.04 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
CCCCCCOC(=O)c1ccccc1O
InChI
1S/C13H18O3/c1-2-3-4-7-10-16-13(15)11-8-5-6-9-12(11)14/h5-6,8-9,14H,2-4,7,10H2,1H3
InChI key
DUKPKQFHJQGTGU-UHFFFAOYSA-N
General description
Hexyl salicylate belongs to the class of flavor & fragrance standards, for use in food, beverage and cosmetic industry. These standards are widely used to add taste and/or smell to products without aroma, to mask unpleasant odors and to maintain stability of original flavor.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Hexyl salicylate may be used as an internal standard for the determination of allergenic ester, benzyl salicylate in propolis specimens using gas chromatography-mass spectrometry (GC?MS), high-performance liquid chromatography (HPLC), and ultraviolet-visible (UV?Vis) spectrophotometry. It may also be used as a standard fragrance material for the quantification of the analyte in wastewater treatment plants using gas chromatography-mass spectrometry (GC?MS).
Other Notes
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Still not finding the right product?
Explore all of our products under Hexyl salicylate
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
flash_point_f
251.6 °F - closed cup
flash_point_c
122 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

