Skip to Content
Merck
All Photos(1)

Key Documents

59370

Sigma-Aldrich

2-Bromopropane

purum, ≥99.0% (GC)

Synonym(s):

Isopropyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHBr
CAS Number:
Molecular Weight:
122.99
Beilstein:
741852
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥99.0% (GC)

refractive index

n20/D 1.425 (lit.)
n20/D 1.425

bp

59 °C (lit.)

mp

−89 °C (lit.)

density

1.31 g/mL at 25 °C (lit.)

SMILES string

CC(C)Br

InChI

1S/C3H7Br/c1-3(2)4/h3H,1-2H3

InChI key

NAMYKGVDVNBCFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Repr. 1A - STOT RE 2 Inhalation

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung-Sun Kang et al.
Reproductive toxicology (Elmsford, N.Y.), 16(2), 151-159 (2002-04-17)
The toxic effects of 2-bromopropane (2-BP) on the reproductive tracts of male F1 offspring from dams exposed to 2-BP during gestation and lactation were investigated. Ten pregnant (sperm-positive) Sprague-Dawley rats per group were exposed sc to 2-BP at 135, 405
Tae Cheon Jeong et al.
Journal of toxicology and environmental health. Part A, 65(5-6), 383-394 (2002-04-09)
Immunotoxic effects of 2-bromopropane were investigated in male Sprague-Dawley rats. The rats were treated orally daily with 2-bromopropane at 100, 330, or 1000 mg/kg for 28 consecutive days. Four days before necropsy, the rats were immunized intravenously with sheep red
Gaku Ichihara
Nihon eiseigaku zasshi. Japanese journal of hygiene, 60(4), 405-410 (2005-12-24)
In 1995, workers in a Korean electronic factory exhibited oligospermia or amenorrhea. We investigated the toxicity of 2-bromopropane used as an alternative to chlorofluorocarbons in the factory in animal experiments, and clarified that exposure to 2-bromopropane depletes spermatogenic cells in
NTP-CERHR Expert Panel Report on the reproductive and developmental toxicity of 2-bromopropane.
Kim Boekelheide et al.
Reproductive toxicology (Elmsford, N.Y.), 18(2), 189-217 (2004-03-17)
Katsumi Ohtani et al.
Industrial health, 42(2), 219-225 (2004-05-07)
Reproductive toxicity of 2-bromopropane (2BP), a substitute for ozone layer-depleting chloro-fluorocarbon, was found among the workers in an electronics factory in Korea in 1995. Furthermore the importance of testicular toxicity has been realized since the problem of endocrine disruptors arose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service