Skip to Content
Merck
All Photos(2)

Documents

56849

Supelco

Fluorene

certified reference material, TraceCERT®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10
CAS Number:
Molecular Weight:
166.22
Beilstein:
1363491
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

298 °C (lit.)

mp

111-114 °C (lit.)

application(s)

environmental

format

neat

SMILES string

C1c2ccccc2-c3ccccc13

InChI

1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2

InChI key

NIHNNTQXNPWCJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

303.8 °F - closed cup

Flash Point(C)

151.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michèle J Régimbald-Krnel et al.
The Journal of organic chemistry, 78(17), 8789-8795 (2013-08-15)
Diphenylcarbene (DPC) generated by high-intensity laser photolysis of diphenyldiazomethane rearranges to fluorene (FL) by two distinct mechanisms as revealed by methyl-group labeling. Thus, excimer laser irradiation of p,p'-dimethyldiphenyldiazomethane generates 3,6-dimethylfluorene (3,6-DMF) and 2,7-dimethylfluorene (2,7-DMF), which were identified by fluorescence measurements
Yoshiaki Amatatsu
The journal of physical chemistry. A, 116(41), 10182-10193 (2012-09-25)
A fluorene-based light-driven molecular rotary motor with constant rotation has been designed by means of ab initio molecular orbital calculations. A model molecule is obtained by a chemical modification of 9-(5-methyl-2-phenyl-2-cyclopenten-1-ylidene)-9H-fluorene (MPCPF) which we reported recently. Despite that MPCPF has
Pierre Lovera et al.
Journal of nanoscience and nanotechnology, 13(7), 5194-5202 (2013-08-02)
Organic nanowires based on a fluorene homopolymer and a copolymer, i.e., poly(9,9-dioctylfluorene), F8, and poly(9,9-dioctylfluorene-co-benzothiadiazole), F8BT, respectively, were synthesised by solution assisted wetting of porous anodic alumina templates. Nanowires ranged between 3 microm and 50 microm in length, and were
S A Schmid et al.
Philosophical transactions. Series A, Mathematical, physical, and engineering sciences, 370(1972), 3787-3801 (2012-07-04)
We have investigated the energy transfer dynamics in a supramolecular linear polymer chain comprising oligofluorene (OF) energy donor units linked by quadruple hydrogen-bonding groups, and oligophenylene (OPV) chain ends that act as energy acceptors. Using femtosecond spectroscopy, we followed the
Qianling Cui et al.
ACS applied materials & interfaces, 5(1), 213-219 (2012-12-15)
In the present work, a facile one-pot method is designed to fabricate a core-shell fluorescent nanoparticle (NP) for cellular imaging based on a new cationic conjugated polymer, poly[9,9'-bis(6,6'-(N,N,N-trimethylaminium)fluorene-2,7-ylenevinylene-co-alt-2,5-dicyano-1,4-phenylene] (PFVCN). Gold nanoflowers (AuNFs) are prepared by a seedless method, in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service