Skip to Content
Merck

Skip To

55876

Adonitol Disks

suitable for microbiology, Sterile filter paper discs impregnated with Adonitol

Synonym(s):

Carbohydrate discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 x 25 discs
Please contact Customer Service for Availability
2 620,00 CZK

About This Item

NACRES:
NA.85
UNSPSC Code:
41171621
Sterility:
sterile
Packaging:
pkg of 10 × 25 discs

2 620,00 CZK


Please contact Customer Service for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 10 × 25 discs

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical, microbiology

storage temp.

2-8°C

suitability

Enterobacter spp., Klebsiella spp., bacteria

General description

Adonitol Discs are used to differentiate bacteria based on the adonitol fermentation ability. Microorganisms can only ferment certain carbohydrates, depending on the enzymes they possess to break down carbohydrates. Fermentation can be detected by gas production (CO2) in liquid media and/or color change of pH caused by acid production. The adonitol discs are aseptically added to any sugar-free basal media to test the ability of the organism to hydrolyze adonitol, crystalline pentose alcohol formed by the reduction of ribose, as the sole source of carbon and energy during fermentation. Adonitol is typically fermented under acid production by many Enterobacter bacteria.

Application

Adonitol Discs are recommended to detect adonitol fermenting bacteria in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
730445648193998
description

suitable for microbiology, Sterile filter paper discs impregnated with Adonitol

description

suitable for microbiology, Sterile filter paper discs impregnated with dulcitol

description

suitable for microbiology, Sterile filter paper discs impregnated with cellobiose

description

suitable for microbiology, Sterile filter paper discs impregnated with sorbitol

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

packaging

pkg of 10 × 25 discs

packaging

pkg of 10 × 25 discs

packaging

pkg of 10 × 25 discs

packaging

pkg of 10 × 25 discs

application(s)

microbiology, clinical testing
environmental
food and beverages
pharmaceutical

application(s)

microbiology, clinical testing
environmental
food and beverages
pharmaceutical

application(s)

microbiology, clinical testing
environmental
food and beverages
pharmaceutical

application(s)

clinical testing
environmental
food and beverages
pharmaceutical, microbiology

technique(s)

microbe id | utilization test: suitable

technique(s)

microbe id | utilization test: suitable

technique(s)

microbe id | utilization test: suitable

technique(s)

microbe id | utilization test: suitable

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters