Skip to Content
Merck

53649

FluoroSELECT E.coli Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
41171621
Suitability:
Escherichia coli, algae, coliforms
Application(s):
food and beverages
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


application(s)

food and beverages

storage temp.

2-8°C

suitability

Escherichia coli, algae, coliforms

General description

The FluoroSELECT product line combines portable, low cost, single-channel fluorometers with suitable kits for a wide range of applications.


Learn more about our FluoroSELECT

Other Notes

Composition (for 50 assays)
Reagent A (Substrate): min. 4 mL
Reagent B (Enzyme Inducer): min. 2 mL
Reagent C (Lysing Agent): min. 6 mL
Incubation Media Powder: 0.4 g
Large Plastic Vials (Sample): 50 pcs
Small Plastic Vials (Mixing): 50 pcs
Mini Glass Tubes (Testing): 50 pcs.

Legal Information

FluoroSELECT is a trademark of Sigma-Aldrich International GmbH


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

104.0 - 122.0 °F

flash_point_c

40 - 50 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library