Skip to Content
Merck
All Photos(2)

Key Documents

52919

Supelco

Hexatriacontane

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CZK 1,330.00

CZK 1,330.00


Please contact Customer Service for Availability
This item is subject to Excise Duty. Please check the cart for more details.

Request a Bulk Order

Select a Size

Change View
250 MG
CZK 1,330.00

About This Item

Linear Formula:
CH3(CH2)34CH3
CAS Number:
Molecular Weight:
506.97
Beilstein:
1712367
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CZK 1,330.00


Please contact Customer Service for Availability
This item is subject to Excise Duty. Please check the cart for more details.

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

17.6 (vs air)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

265 °C/1 mmHg (lit.)

mp

74-76 °C (lit.)
75-77 °C

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C36H74/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-36H2,1-2H3

InChI key

YDLYQMBWCWFRAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Hexene can undergo bio-oxidation to form 5-hexen-1-ol, which can undergo further oxidation to yield 5-hexenoic acid.[1] It can also find applications as a comonomer in the production of polyethylene.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Montgomery.HJ
Groundwater Chemicals Desk Reference, (4) (2007)
Klerk DA
Fischer-Tropsch Refining (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service