Skip to Content
Merck

Skip To

52203

Methyl hexacosanoate

analytical standard

Synonym(s):

Hexacosanoic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
CZK 7,640.00

About This Item

Linear Formula:
CH3(CH2)24COOCH3
CAS Number:
Molecular Weight:
410.72
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
227-355-6
Beilstein/REAXYS Number:
1800770
MDL number:

CZK 7,640.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

63-66 °C

format

neat

functional group

ester

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C27H54O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27(28)29-2/h3-26H2,1-2H3

InChI key

VHUJBYYFFWDLNM-UHFFFAOYSA-N

General description

Methyl hexacosanoate is a compound, which can be isolated from lingwu bituminous coal.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H63895153587115
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

format

-

format

neat

format

neat

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

grade

analytical standard

grade

-

grade

analytical standard

grade

analytical standard

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

assay

≥99.0% (GC)

assay

≥99% (capillary GC)

assay

≥98.5% (GC)

assay

≥99.0% (GC)


Still not finding the right product?

Explore all of our products under Methyl hexacosanoate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters