Skip to Content
Merck
All Photos(1)

Key Documents

51962

Millipore

m-CP selective Supplement I

suitable for microbiology

Synonym(s):

Membrane Clostridium Perfringens Selective Supplement I, Polymyxin B sulfate - D-cycloserine mixture

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
CZK 2,160.00

CZK 2,160.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 VIALS
CZK 2,160.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

CZK 2,160.00


Please contact Customer Service for Availability

Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Clostridium spp.

General description

An antibiotic supplement recommended by the Directive of the Council of the European Union 98/83/EC for the selective isolation of Clostridium perfringens.

Application

for the selective isolation of Clostridium perfringens

Components

(One vial is sufficient for 500 mL medium)
D-Cycloserine 200.0 mg, Polymyxin B sulphate 12.5 mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service