Skip to Content
Merck

Skip To

46256

Erythromycin A hydrate

VETRANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mg
Please contact Customer Service for Availability
CZK 1,840.00

About This Item

Empirical Formula (Hill Notation):
C37H67NO13 · xH2O
CAS Number:
Molecular Weight:
733.93 (anhydrous basis)
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
EC Number:
204-040-1
NACRES:
NA.24

CZK 1,840.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

N([C@@H]1[C@H]([C@@H](O[C@@H](C1)C)O[C@@H]2[C@H]([C@@H]([C@H](C(=O)O[C@@H]([C@]([C@@H]([C@H](C(=O)C(C[C@]2(O)C)C)C)O)(O)C)CC)C)O[C@@H]3O[C@H]([C@@H]([C@@](C3)(OC)C)O)C)C)O)(C)C.O.O

InChI

1S/C37H67NO13.2H2O/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;;/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;2*1H2/t18?,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-;;/m1../s1

InChI key

IWGQNYZQLVGGCS-ZKSNWDMWSA-N

Application

Erythromycin is an antibiotic produced by growth of certain strains of Streptomyces erythreus. This product is composed largely of erythromycin A with small amounts of erythromycins B and C and is recommended for concentration at 100 mg/L. Concentrations between 50 and 200 mg/L have also proven effective in controlling bacterial growth. Erythromycin has been used as a motilin receptor agonist, to block respiratory glycoconjugate secretion in human airways in vitro, and for selecting plasmid-cured and recombinant lactococcus lactis MG1363 strains.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Mode of Action: Erythromycin acts by inhibiting elongation at the transpeptidation step, specifically aminoacyl translocation from the A-site to P-site.

Antimicrobial Spectrum: This product acts against both gram-negative and gram-positive bacteria.

Preparation Note

This product is soluble in water at 2 mg/mL, with a 0.067% solution in water yielding a pH of 8.0-10.5. It is also soluble in ethanol at 50 mg/mL, yielding a clear, colorless to faint yellow solution. It is freely soluble in alcohol, acetone, chloroform, acetonitrile and ethyl acetate but forms salts with acids. All solutions should be protected from light.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is stable in solution at 37°C for 3 days. Stock solutions should be stored at 2-8°C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
329044682646589
application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters