Skip to Content
Merck
All Photos(1)

Documents

46019

Supelco

Terbuthylazine-2-hydroxy

PESTANAL®, analytical standard

Synonym(s):

Hydroxyterbuthylazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17N5O
CAS Number:
Molecular Weight:
211.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(O)nc(NC(C)(C)C)n1

InChI

1S/C9H17N5O/c1-5-10-6-11-7(13-8(15)12-6)14-9(2,3)4/h5H2,1-4H3,(H3,10,11,12,13,14,15)

InChI key

OYTCZOJKXCTBHG-UHFFFAOYSA-N

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023
Terbutylazine-2-hydroxy is a metabolite of triazine herbicide. It is used for the pre- and posr-emergence control of annual weeds, perennial weeds, grasses, and broadleaf weeds in agricultural crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support. Terbutylazine-2-hydroxy may be used as an analytical reference standard for the quantification of the analyte in environmental samples using various chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Velisek J, et al.
BioMed Research International (2014)
Sebastian Huntscha et al.
Journal of chromatography. A, 1268, 74-83 (2012-11-10)
An automated multiresidue method consisting of an online solid-phase extraction step coupled to a high performance liquid chromatography-tandem mass spectrometer (online-SPE-HPLC-MS/MS method) was developed for the determination of 88 polar organic micropollutants with a broad range of physicochemical properties (logD(OW)
P Serra-Mora et al.
Journal of chromatography. A, 1513, 51-58 (2017-07-30)
In-tube solid-phase microextraction (IT-SPME) coupled to miniaturized liquid chromatography (LC) techniques are attractive mainly due to the column efficiency improvement, sensitivity enhancement and reduction of solvent consumption. In addition, the nanomaterials based sorbents can play a key role in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service