Skip to Content
Merck
All Photos(1)

Key Documents

45898

Supelco

Propham solution

100 μg/mL in methanol, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
2209666
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CC(C)OC(=O)Nc1ccccc1

InChI

1S/C10H13NO2/c1-8(2)13-10(12)11-9-6-4-3-5-7-9/h3-8H,1-2H3,(H,11,12)

InChI key

VXPLXMJHHKHSOA-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Corti et al.
Food additives and contaminants, 8(5), 607-615 (1991-09-01)
HPTLC was used to check for residues of chlorpropham, propham (sprout inhibitors) and thiabendazole (fungicide) applied to potatoes. The method used gave good precision and analytical sensitivity. The sample preparation method developed by the authors was simple and gave good
Microtubule organizing centers (MTOCs) of the alga Polytomella exert spatial control over microtubule initiation in vivo and in vitro.
M E Stearns et al.
Journal of ultrastructure research, 77(3), 366-378 (1981-12-01)
M A Pilinskaia
TSitologiia i genetika, 20(2), 143-145 (1986-03-01)
The cytogenetic activity of some substances formed in agricultural plants during metabolism of pesticides of four classes of chemical compounds was studied in the culture of human peripheric blood lymphocytes. Metabolites were shown either to have mutagenic properties similar to
R Michel et al.
Comptes rendus des seances de la Societe de biologie et de ses filiales, 174(2), 176-183 (1980-01-01)
Propham and Chloropropham disturb the several mitotic figure components of proliferating human lymphocytes. The tubular structures, affected in preference by the C.I.P.C., become abnormal, disorganized or disappear. The chromosomal changes (protuberances,absence of condensation and chromosomal bridges) are principally given by
X Pous et al.
Fresenius' journal of analytical chemistry, 371(2), 182-189 (2001-10-27)
Imidacloprid, metalaxyl, myclobutanil, propham, and thiabendazole have been simultaneously determined in strawberries, oranges, potatoes, pears, and melons by matrix solid-phase dispersion (MSPD) followed by liquid chromatography-atmospheric pressure chemical ionization-mass spectrometry (LC-APCI-MS) in positive-ion mode. The samples were homogenized with C8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service