Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
ClCF2CCl2F
CAS Number:
Molecular Weight:
187.38
EC Number:
200-936-1
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
1740335
MDL number:
grade
analytical standard
vapor pressure
5.5 psi ( 20 °C)
assay
≥99.7% (GC)
autoignition temp.
1256 °F
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
refractive index
n
bp
47-48 °C (lit.)
mp
−35 °C (lit.)
density
1.57 g/mL at 25 °C (lit.)
application(s)
environmental
SMILES string
FC(F)(Cl)C(F)(Cl)Cl
InChI
1S/C2Cl3F3/c3-1(4,6)2(5,7)8
InChI key
AJDIZQLSFPQPEY-UHFFFAOYSA-N
Legal Information
Genetron is a registered trademark of Honeywell International Inc.
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
383.0 °F
flash_point_c
195 °C
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

