Skip to Content
Merck
All Photos(1)

Key Documents

44394

Supelco

Profenofos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 4,540.00

CZK 4,540.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CZK 4,540.00

About This Item

Empirical Formula (Hill Notation):
C11H15BrClO3PS
CAS Number:
Molecular Weight:
373.63
Beilstein:
2150258
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 4,540.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

CCCSP(=O)(OCC)Oc1ccc(Br)cc1Cl

InChI

1S/C11H15BrClO3PS/c1-3-7-18-17(14,15-4-2)16-11-6-5-9(12)8-10(11)13/h5-6,8H,3-4,7H2,1-2H3

InChI key

QYMMJNLHFKGANY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Profenofos is a non-systemic insecticide and acaricide that belongs to the group of organophosphates. It is an acetylcholinesterase (AChE) inhibitor causing hyperexcitation and impedes the functioning of the nervous system. Profenofos is used for protection against insects, particularly Lepidoptera, and mites in cotton, maize, sugar beets, soybeans, potatoes, vegetables, tobacco, and other crops.

According to the Commission Implementing Regulation (EU) No 2076/2002 of 20th November 2002, profenofos is not approved for use in the European Union (EU) as a plant protection agent, following Regulation (EC) No. 1107/2009. But maximum residue levels of 0.01 mg/kg to 10 mg/kg are allowed for the presence of profenofos in products of plant and animal origin, according to the EU Reg. No 2017/978.
Profenofos has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Profenofos CRM may also be used as given below:

  • Evaluation of Fourier transform-NIR (FT-NIR) and Fourier transform-MIR (FT-MIR) spectroscopy techniques for the detection of profenofos in Chinese kale, cabbage, and chili spur pepper samples
  • Multi-residue analysis of pesticides in 150 egg samples comprising duck, goose, and quail eggs by a modified QuEChERS method for extraction and liquid chromatography-tandem mass spectrometry (LC-MS/MS) for quantification
  • Detection of insecticides from three different classes in peach samples by surface-enhanced Raman scattering (SERS) technique, based on silver-coated gold nanoparticles (Au@Ag NPs)
  • Multi-residue analysis of 160 multiclass pesticides using ethyl acetate (EtAC)/n. hexane mixture for extraction from chamomile, thyme, and marjoram samples and their analysis by gas chromatography-tandem mass spectrometry (GC-MS/MS)
  • Simultaneous determination of 18 chiral pesticides in solid and liquid environmental samples by magnetic amino-functionalized multiwalled carbon nanotubes (m-MWCNTs-NH2) based extraction prior to an enantioselective liquid chromatography-tandem mass spectrometric (LC-MS/MS) analysis
  • Method development and validation for the determination of 36 multi-class pesticides in soil samples by liquid chromatography with tandem quadrupole linear ion trap mass spectrometry (MS-MS/MS) following a modified QuEChERS-based extraction

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Feng et al.
Journal of separation science, 38(17), 3047-3054 (2015-06-23)
A new method for simultaneous determination of 36 pesticides, including 15 organophosphorus, six carbamate, and some other pesticides in soil was developed by liquid chromatography with tandem quadruple linear ion trap mass spectrometry. The extraction and clean-up steps were optimized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service