Skip to Content
Merck

Skip To

44022

KPC ChromoSelect Agar Base

NutriSelect® Plus, suitable for microbiology

Synonym(s):

HiCrome ′Klebsiella pneumoniae carbapenemase′ Agar Base, HiCrome ′carbapenemase resistant Enterobacteriaceae′ Agar Base, HiCrome CRE Agar Base

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 g
Please contact Customer Service for Availability
CZK 8,280.00

About This Item

NACRES:
NA.85
UNSPSC Code:
41171607

CZK 8,280.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L , chromogenic mixture, 3.0 g/L , peptone special, 15.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.0±0.2 (25 °C)

application(s)

clinical testing
veterinary, microbiology

storage temp.

2-8°C

suitability

Klebsiella spp.

Application

KPC ChromoSelect Agar Base is recommended for the detection of gram negative bacteria with a reduced susceptibility to a carbapenem agents.

Preparation Note

Suspend 16.5 grams in 500 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool to 45-50°C. Aseptically add rehydrated contents of 1 vial of KPC ChromoSelect Selective Supplement (Cat. No. 52099). Mix well and pour into sterile Petri plates.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
HiCrome is a trademark of HiMedia Laboratories Pvt. Ltd.
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
909259232492325
suitability

Klebsiella spp.

suitability

selective for Klebsiella spp.

suitability

selective and differential for Cronobacter spp., selective and differential for Enterobacter spp.

suitability

selective for Bacillus cereus

form

powder

form

powder

form

powder

form

powder

composition

agar, 15.0 g/L , peptone special, 15.0 g/L , chromogenic mixture, 3.0 g/L

composition

agar, 15.0 g/L , bile salts mixture, 1.5 g/L , chromogenic mixture, 0.2 g/L , petone, special, 12.0 g/L , sodium chloride, 5.0 g/L , sodium lauryl sulfate, 0.1 g/L , yeast extract, 7.0 g/L

composition

agar, 15.0 g/L , casein enzymic hydrolysate, 15.0 g/L , chromogenic mixture, 10.17 g/L , papaic digest of soyabean meal, 5.0 g/L , sodium chloride, 5.0 g/L , sodium deoxycholate, 0.5 g/L , sodium thiosulfate, 1.0 g/L

composition

agar, 15.0 g/L , chromogenic mixture, 3.2 g/L , D-mannitol, 10.0 g/L , meat extract, 1.0 g/L , peptic digest of animal tissue, 10 g/L , phenol red, 0.025 g/L , sodium chloride, 10.0 g/L

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

application(s)

clinical testing
veterinary, microbiology

application(s)

bioburden testing
clinical testing
environmental
food and beverages
water monitoring, microbiology

application(s)

clinical testing
environmental
food and beverages
veterinary, microbiology

application(s)

clinical testing
environmental
food and beverages, microbiology

final pH

7.0±0.2 (25 °C)

final pH

7.1±0.2 (25 °C)

final pH

7.3±0.2 (25 °C)

final pH

7.1±0.2 (25)


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters