Skip to Content
Merck

43729

1-Methyl-2-pyrrolidinone

for metal speciation analysis, ≥99.0% (GC)

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
UNSPSC Code:
41116105
NACRES:
NA.24
PubChem Substance ID:
EC Number:
212-828-1
Beilstein/REAXYS Number:
106420
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C), 0.99 mmHg ( 40 °C)

assay

≥99.0% (GC)

autoignition temp.

518 °F

quality

for metal speciation analysis

expl. lim.

9.5 %

impurities

≤0.1% water

refractive index

n20/D 1.47 (lit.), n20/D 1.470

bp

202 °C (lit.), 81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤100 μg/kg, chloride (Cl-): ≤100 μg/kg, iodide (I-): ≤100 μg/kg, nitrate (NO3-): ≤200 μg/kg, nitrite (NO2-): ≤100 μg/kg, phosphate (PO43-): ≤100 μg/kg, sulfate (SO42-): ≤1000 μg/kg

SMILES string

CN1CCCC1=O

cation traces

Ag: ≤2 μg/kg, Al: ≤5 μg/kg, As: ≤0.5 μg/kg, Au: ≤0.5 μg/kg, Ba: ≤2 μg/kg, Be: ≤0.5 μg/kg, Bi: ≤0.5 μg/kg, Ca: ≤10 μg/kg, Cd: ≤0.5 μg/kg, Ce: ≤0.5 μg/kg, Co: ≤0.5 μg/kg, Cr: ≤1 μg/kg, Cs: ≤0.5 μg/kg, Cu: ≤5 μg/kg, Fe: ≤10 μg/kg, Ga: ≤0.5 μg/kg, Ge: ≤1 μg/kg, Hg: ≤1 μg/kg, In: ≤0.5 μg/kg, Ir: ≤0.5 μg/kg, K: ≤20 μg/kg, Li: ≤0.5 μg/kg, Mg: ≤1 μg/kg, Mn: ≤0.5 μg/kg, Mo: ≤0.5 μg/kg, Na: ≤500 μg/kg, Ni: ≤1 μg/kg, Pb: ≤0.5 μg/kg, Pd: ≤0.5 μg/kg, Pt: ≤0.5 μg/kg, Rb: ≤1 μg/kg, Rh: ≤0.5 μg/kg, Ru: ≤0.5 μg/kg, Sb: ≤0.5 μg/kg, Se: ≤5 μg/kg, Sn: ≤0.5 μg/kg, Sr: ≤0.5 μg/kg, Ti: ≤1 μg/kg, Tl: ≤0.5 μg/kg, V: ≤1 μg/kg, Zn: ≤10 μg/kg, Zr: ≤1 μg/kg

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

General description

NMP is an effective solvent for organic and inorganic compounds. Tertiary amides in H-bond basicity are decreased by bulky substituents on carbon than bulky substituents on nitrogen. It is also an effective solvent for the decarbonylation of aldehydes. It exhibits as reagent and amidines formed from NMP exist as amidine-enediamine tautomers. As cosolvent it has ability to enhance the reactivity of other reagents.

Application

NMP was used:
  • as an aqueous physical solvent to measure the molar heat capacities.
  • in the synthesis of Nafion/SiO2 nanocomposite membranes.
  • in the synthesis of N-Butyl-N,N-dimethyl chitosan iodide (QCh).
  • as an inducer to demonstrate phenotypic resistance to Chloramphenicol and ampicillin in Escherichia coli K-12 starins.
  • used as polar solvent inducer for erythroleukemic cell differentiation.
  • as a blocking reagent in microarray manufacturing.


Still not finding the right product?

Explore all of our products under 1-Methyl-2-pyrrolidinone


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

195.8 °F - Pensky-Martens closed cup

flash_point_c

91 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

872-50-4

CAS No.

872-50-4

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library