Skip to Content
Merck

Skip To

428604

AmberChrom 1X4 Ion Exchange Resin

chloride form, anion, 200-400 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 3,320.00
500 g
Please contact Customer Service for Availability
CZK 12,500.00

About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:

CZK 3,320.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberChrom 1X4 chloride form, 200-400 mesh, Anion Exchange Resin

Quality Segment

description

cross-linkage 4%

form

beads

parameter

66 °C OH- form max. temp., 99 °C Cl- form max. temp.

moisture

55-63%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

200-400 mesh

operating pH

0-14

capacity

1.0 meq/mL capacity

separation technique

anion exchange

SMILES string

N(C)(C)C.C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

Application

Strongly basic (type I) anion exchange resin for fine pharmaceuticals; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.
This 1X4, anion-exchange resin, was used as adsorbent material in characterization of iodinated contrast media using 11B NMR and capillary electrophoresis.[1] The unreacted fluoride in a solution was removed by passing the solution through a column packed with 1X4-50 chloride form.[2]

Features and Benefits

strongly basic anion.

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
428612428590217395
parameter

66 °C OH- form max. temp., 99 °C Cl- form max. temp.

parameter

66 °C OH- form max. temp., 99 °C Cl- form max. temp.

parameter

66 °C OH- form max. temp., 99 °C Cl- form max. temp.

parameter

66 °C OH- form max. temp., 99 °C Cl- form max. temp.

matrix

styrene-divinylbenzene (gel)

matrix

styrene-divinylbenzene (gel)

matrix

styrene-divinylbenzene (gel)

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

matrix active group

quaternary ammonium

matrix active group

quaternary ammonium

matrix active group

trimethylbenzylammonium functional group

separation technique

anion exchange

separation technique

anion exchange

separation technique

anion exchange

separation technique

anion exchange

particle size

200-400 mesh

particle size

20-50 mesh

particle size

100-200 mesh

particle size

200-400 mesh

form

beads

form

beads

form

beads

form

beads


Still not finding the right product?

Explore all of our products under AmberChrom 1X4 chloride form


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. I am looking for information about true density of AmberChrom™ 1X4 Ion Exchange Resin chloride form, anion, 200-400 mesh, COA MKCP1981

    1 answer
    1. Unfortunately, the density of this product has not been determined.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. How can a Dowex type 1 resin in the chloride form be converted to the carbonate form?

    1 answer
    1. As one might expect, the procedure requires the use of sodium carbonate (Na2CO3).Citation: Journal of Experimental Medicine, 120, 1007-1018 (1965).From page 1011 of that paper:"A 2 mL column of washed Dowex-1X8 200 to 400 mesh, in the chloride form, was converted to the carbonate form by washing with 1 M Na2CO3 until no chloride could be detected in the eluate. It was then washed with distilled water until the pH of the eluate reached neutrality". The simplest test for chloride is to use a dilute silver nitrate solution. If mixing a few drops of the eluate with a few drops of a silver nitrate solution results in a silver chloride precipitate forming, that would an indication that the eluate contains chloride. You could also use a chloride-specific electrode, if you have access to one.

      Helpful?

Reviews

No rating value

Active Filters