Skip to Content
Merck
All Photos(2)

Key Documents

42304

Supelco

Isophthalic acid

analytical standard

Synonym(s):

Benzene-1,3-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein:
1909332
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

autoignition temp.

1198 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

341-343 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(=O)c1cccc(c1)C(O)=O

InChI

1S/C8H6O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

QQVIHTHCMHWDBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Isophthalic acid may be used as a reference standard in the analysis of isophthalic acid from PET bottles into edible oils using high performance liquid chromatography (HPLC) technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC study of migration of terephthalic acid and isophthalic acid from PET bottles into edible oils.
Mousavi K, et al.
Journal of the Science of Food and Agriculture, 94(11), 2205-2209 (2014)
Yucui Hou et al.
The journal of physical chemistry. B, 118(47), 13646-13650 (2014-10-30)
Because of similar properties and very low volatility, isomers of benzene poly(carboxylic acid)s (BPCAs) are very difficult to separate. In this work, we found that isomers of BPCAs could be separated efficiently by quaternary ammonium salts (QASs) via formation of
Dan Song et al.
Journal of neuroscience research, 93(1), 130-139 (2014-08-16)
The cotransporter of Na(+) , K(+) , 2Cl(-) , and water, NKKC1, is activated under two conditions in the brain, exposure to highly elevated extracellular K(+) concentrations, causing astrocytic swelling, and regulatory volume increase in cells shrunk in response to
Kerstin Pohl et al.
Blood, 124(7), 999-1009 (2014-06-18)
Studies have endeavored to reconcile whether dysfunction of neutrophils in people with cystic fibrosis (CF) is a result of the genetic defect or is secondary due to infection and inflammation. In this study, we illustrate that disrupted function of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service