Skip to Content
Merck
All Photos(1)

Key Documents

42036

Sigma-Aldrich

3,5-Dinitrobenzyl chloride

purum, ≥97.0% (AT)

Synonym(s):

α-Chloro-3,5-dinitrotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CZK 3,370.00
25 G
CZK 11,600.00

CZK 3,370.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CZK 3,370.00
25 G
CZK 11,600.00

About This Item

Linear Formula:
(O2N)2C6H3CH2Cl
CAS Number:
Molecular Weight:
216.58
Beilstein:
2506587
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 3,370.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

purum

Quality Level

Assay

≥97.0% (AT)

mp

79-82 °C (lit.)

functional group

chloro
nitro

SMILES string

[O-][N+](=O)c1cc(CCl)cc(c1)[N+]([O-])=O

InChI

1S/C7H5ClN2O4/c8-4-5-1-6(9(11)12)3-7(2-5)10(13)14/h1-3H,4H2

InChI key

SMJODKZAFKWUJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dinitrobenzyl chloride was used in the synthesis of double-spanned double cavity calix[4]arenes.[1] It was used in the fuorescence flow immunoassay for the detection of 4-nitrophenol.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calixarenes. 27. Synthesis, characterization, and complexation studies of double-cavity calix [4] arenes.
Gutsche CD and See KA.
The Journal of Organic Chemistry, 57(16), 4527-4539 (1992)
Competitive flow immunoassay with fluorescence detection for determination of 4-nitrophenol.
Nistor C, et al.
Analytica Chimica Acta, 426(2), 185-195 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service