Skip to Content
Merck

Skip To

41932

L-Lysine decarboxylation medium

NutriSelect® Plus, powder, for enterobacteriaceae, pack of 500 g, suitable for microbiology

Synonym(s):

LD medium, Lysine Decarboxylase Broth without Peptone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 g
Please contact Customer Service for Availability
CZK 6,400.00

About This Item

NACRES:
NA.74
UNSPSC Code:
41171606
Manufacturer/tradename:
NutriSelect® Plus
Agency:
according to GB 4789.40-2016, according to ISO 19250:2010, according to ISO 22964:2017
Form:
powder
Suitability:
Salmonella spp., enterobacteriaceae
Application(s):
environmental
food and beverages, microbiology

CZK 6,400.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

according to GB 4789.40-2016, according to ISO 19250:2010, according to ISO 22964:2017

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bromocresol purple, 0.015 g/L , dextrose, 1.0 g/L , L-lysine hydrochloride, 5.0 g/L , yeast extract, 3.0 g/L

packaging

pack of 500 g

manufacturer/tradename

NutriSelect® Plus

final pH

6.8±0.2 (25 °C)

application(s)

environmental
food and beverages, microbiology

suitability

Salmonella spp., enterobacteriaceae

Application

Lysine Decarboxylase Broth w/o Peptone are used for differentiating Salmonella arizonae from the Bethesda Ballerup group of Enterobacteriaceae.

Preparation Note

Suspend 9.01 grams in 1000 ml distilled water. Heat if necessary to dissolve the medium completely. Dispense 5 ml amount into screw-capped test tubes. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
663041478122098
agency

according to GB 4789.40-2016, according to ISO 22964:2017, according to ISO 19250:2010

agency

according to ISO 10273

agency

according to ISO 6579-1:2017

agency

according to ISO 10273:2017, according to ISO 21871:2006

application(s)

environmental
food and beverages, microbiology

application(s)

clinical testing
environmental
food and beverages, microbiology

application(s)

food and beverages
water monitoring, microbiology

application(s)

bioburden testing
cleaning products
clinical testing
cosmetics
environmental
food and beverages
pathogen testing
personal care
pharmaceutical
sample preparation
veterinary
water monitoring, microbiology

form

powder

form

powder

form

powder

form

powder

suitability

Salmonella spp., enterobacteriaceae

suitability

Enterobacter spp., Salmonella spp., enterobacteriaceae

suitability

Salmonella spp.

suitability

bacteria (fastidious organism), nonselective for Bacillus spp., nonselective for Bacteroides spp., nonselective for Campylobacter spp., nonselective for Candida spp., nonselective for Escherichia coli, nonselective for Neisseria spp., nonselective for Staphylococcus spp., nonselective for Streptococcus spp., nonselective for coliforms

manufacturer/tradename

NutriSelect® Plus

manufacturer/tradename

NutriSelect® Plus

manufacturer/tradename

NutriSelect® Plus

manufacturer/tradename

NutriSelect® Basic

composition

bromocresol purple, 0.015 g/L , L-lysine hydrochloride, 5.0 g/L , dextrose, 1.0 g/L , yeast extract, 3.0 g/L

composition

bromo creol purple, 0.02 g/L , dextrose, 1 g/L , L-lysine hydrochloride, 5 g/L , peptic digest of animal tissue, 5 g/L , yeast extract, 3 g/L

composition

agar, 15.0 g/L , ferric ammonium citrate, 0.80 g/L , lactose, 7.5 g/L , L-lysine hydrochloride, 5.0 g/L , phenol red, 0.08 g/L , sodium chloride, 5.0 g/L , sodium deoxycholate, 1.0 g/L , sodium thiosulfate, 6.8 g/L , sucrose, 7.5 g/L , xylose, 3.75 g/L , yeast extract, 3.0 g/L

composition

casein peptone, 17 g/L , dipotassium hydrogen phosphate, 2.5 g/L , D(+)-glucose, 2.5 g/L , sodium chloride, 5 g/L , soy peptone, 3 g/L


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters