Skip to Content
Merck

Skip To

41724

DL-threo-Chloramphenicol-d5

analytical standard

Synonym(s):

DL-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-β-(4-nitrophenyl-2,3,5,6-d4)ethyl-β-d]acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Please contact Customer Service for Availability
CZK 11,800.00

About This Item

Empirical Formula (Hill Notation):
C11D5H7Cl2N2O5
Molecular Weight:
328.16
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 11,800.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+5

SMILES string

[2H]c1c([2H])c(c([2H])c([2H])c1[N+]([O-])=O)C([2H])(O)C(CO)NC(=O)C(Cl)Cl

InChI

1S/C11H12Cl2N2O5/c12-10(13)11(18)14-8(5-16)9(17)6-1-3-7(4-2-6)15(19)20/h1-4,8-10,16-17H,5H2,(H,14,18)/i1D,2D,3D,4D,9D

InChI key

WIIZWVCIJKGZOK-NEKQGQCZSA-N

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706024

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
493585378753541
application(s)

clinical testing

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, solid phase extraction (SPE): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, solid phase extraction (SPE): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, solid phase extraction (SPE): suitable

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

assay

≥97.0% (HPLC)

assay

≥98.5% (HPLC)

assay

≥98.5% (HPLC)

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters