Skip to Content
Merck

40925

Chloro-dimethyl(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane

purum, ≥97.0% (GC)

Synonym(s):

3,3,4,4,5,5,6,6,6-Nonafluorohexyl-dimethylchlorosilane, Chloro-dimethyl(1H,1H,2H,2H-perfluorhexyl)silane, DMNFHSCl, Dimethyl(3,3,4,4,5,5,6,6,6-nonafluorohexyl)chlorosilane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H10ClF9Si
CAS Number:
Molecular Weight:
340.69
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
2112822
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (GC)

refractive index

n20/D 1.350

bp

159-161 °C (lit.)

density

1.360 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Other Notes

Derivatizing agent used for the determination of compounds in picomolar amounts by gas chromatogr. - selected ion monitoring mass spectrometry, GC-SIMS, e.g. of juvenile hormones




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library