Skip to Content
Merck

Skip To

40733

trans-2-Tetradecenoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[[(2E)-1-oxo-2-tetradecen-1-yl]oxy]-1-propanaminium inner salt, trans-2-Myristenoyl-L-carnitine, C14:1-Carnitine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
CZK 3,700.00

About This Item

Empirical Formula (Hill Notation):
C21H39NO4
Molecular Weight:
369.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 3,700.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

assay

≥95.0% (LC/MS)

optical activity

[α]/D -12±2°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

[O-]C(C([H])([H])[C@](OC(/C([H])=C([H])/C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])[H])=O)([H])C([H])([H])[N+](C([H])([H])[H])(C([H])([H])[H])C([H])([H])[H])=O

InChI

1S/C21H39NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4/h15-16,19H,5-14,17-18H2,1-4H3/b16-15+/t19-/m1/s1

InChI key

BUBHHSHBKWNLLG-CLHDZIPGSA-N

General description

trans-2-Tetradecenoyl-L-carnitine, belonging to the class of acylcarnitines, serves as a specific diagnostic metabolite biomarker, which increases in very long-chain acyl CoA dehydrogenase (VLCAD) deficiency, existing as a long-chain β-oxidation defect in newborns.

Application

trans-2-Tetradecenoyl-L-carnitine may be used as an analytical reference standard in the following:
  • Quantification of the analyte in biological samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC/MS/MS).
  • Quantification of the analyte in dried blood spots using chip-based nanoelectrospray source coupled with high resolution mass spectrometry (chip-based nanoESI- HRMS) and targeted liquid chromatography-tandem mass spectrometry (LC/MS/MS).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
915219138830172
format

neat

format

neat

format

neat

format

neat

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

optical activity

[α]/D -12±2°, c = 0.1 in methanol

optical activity

[α]/D -15±3°, c = 0.1 in H2O

optical activity

[α]/D -21.0±2.0°, c = 0.1 in H2O

optical activity

[α]/D -13±3°, c = 0.1 in methanol

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. CAS: 51534-36-2 is that the same trans-2-Tetradecenoyl-L-carnitine?

    1 answer
    1. CAS 51534-36-2 is 2-Tetradecenal with a formula of C14H26O, which is not a product offered. This material is trans-2-tetradecenoyl-L-carnitine, with a formula of C21H39NO4 and the structure shown at the top of the product page.

      Helpful?

Reviews

No rating value

Active Filters