Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3NHNHCH3 · 2HCl
CAS Number:
Molecular Weight:
133.02
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
206-183-5
MDL number:
Beilstein/REAXYS Number:
3905684
grade
puriss.
assay
≥99.0% (AT)
mp
~168 °C (dec.)
SMILES string
Cl.Cl.CNNC
Other Notes
Mutagen, used for the experimental induction of colon cancer
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.