Skip to Content
Merck

39367

1-Dansylpiperazine

≥99.0%

Synonym(s):

1-[5-(Dimethylamino)naphthalene-1-sulfonyl]piperazine

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C16H21N3O2S
CAS Number:
Molecular Weight:
319.42
UNSPSC Code:
41116105
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

derivatization grade ((HPLC))

assay

≥99.0% (HPLC), ≥99.0%

mp

130-135 °C

fluorescence

λex 340 nm; λem 523 nm in methanol

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)N3CCNCC3

InChI

1S/C16H21N3O2S/c1-18(2)15-7-3-6-14-13(15)5-4-8-16(14)22(20,21)19-11-9-17-10-12-19/h3-8,17H,9-12H2,1-2H3

InChI key

NRTFFCBHBQNURK-UHFFFAOYSA-N

Other Notes

Reagent for the fluorimetric determination of fatty acids in body fluids; fatty acids determination by pre-column derivatization

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Junker-Buchheit, H. Jork
Fresenius Journal of Analytical Chemistry, 331, 387-387 (1988)
I Yanagisawa et al.
Journal of chromatography, 345(2), 229-240 (1985-12-13)
The quantitative determination of saturated and unsaturated fatty acids (ranging from acetic acid to lignoceric acid) in biological samples is presented. The secondary amine group of 5-(dimethylamino)-1-naphthalenesulponyl-semipiperazide (dansyl-semipiperazide) reacts with the carboxyl group of the fatty acids to form an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service