Skip to Content
Merck

Skip To

39269

Linoleic acid

Synonym(s):

cis-9,cis-12-Octadecadienoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Please contact Customer Service for Availability
CZK 12,400.00
100 g
Please contact Customer Service for Availability
CZK 68,500.00
1 kg
Please contact Customer Service for Availability
CZK 378,500.00

About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
280.45
EC Number:
200-470-9
UNSPSC Code:
12352211
MDL number:
Beilstein/REAXYS Number:
1727101

CZK 12,400.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.0% (GC)

Quality Segment

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

heavy metals, tested

refractive index

n20/D 1.466 (lit.)

bp

229-230 °C/16 mmHg (lit.)

mp

−5 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(CCCCCCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-

InChI key

OYHQOLUKZRVURQ-HZJYTTRNSA-N

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Biochem/physiol Actions

Linoleic acid increases cell proliferation and gene expression of PPARα and its target genes such as acyl-CoA oxidase in primary duck hepatocytes .

Analysis Note

microbiol contamination ≤ 1000 CFU/g
residual solvents (GLC-HS) conforms ICHQ3

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W800075L1376W338020
form

liquid

form

-

form

liquid

form

-

assay

≥98.0% (GC)

assay

≥95%

assay

≥98%

assay

≥95%

impurities

heavy metals, tested

impurities

-

impurities

-

impurities

-

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

refractive index

n20/D 1.466 (lit.)

refractive index

n20/D 1.466 (lit.)

refractive index

n20/D 1.466 (lit.)

refractive index

n20/D 1.466 (lit.)

bp

229-230 °C/16 mmHg (lit.)

bp

229-230 °C/16 mmHg (lit.)

bp

229-230 °C/16 mmHg (lit.)

bp

229-230 °C/16 mmHg (lit.)


Still not finding the right product?

Explore all of our products under Linoleic acid


wgk

WGK 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters