Skip to Content
Merck

Skip To

390410

3,3′-Diethylthiacyanine iodide

Dye content ~97 %

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 9,460.00

About This Item

Empirical Formula (Hill Notation):
C19H19IN2S2
CAS Number:
Molecular Weight:
466.40
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
218-593-1
MDL number:

CZK 9,460.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥96.5% (HPLC)

Quality Segment

form

powder or crystals

composition

Dye content, ~97%

mp

>300 °C (lit.)

λmax

424 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[I-].CCN1\C(Sc2ccccc12)=C\c3sc4ccccc4[n+]3CC

InChI

1S/C19H19N2S2.HI/c1-3-20-14-9-5-7-11-16(14)22-18(20)13-19-21(4-2)15-10-6-8-12-17(15)23-19;/h5-13H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

XWUVSDMFUQTKQC-UHFFFAOYSA-M

General description

3,3′-Diethylthiacyanine iodide (DTTHCI) is a fluorophore.

Application

3,3′-Diethylthiacyanine iodide has been used to investigate the kinetics of fluorescence staining in bacteria (two Gram-negative and two Gram-positive species).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
318434318426M44451
form

powder or crystals

form

crystalline powder

form

powder or chunks

form

powder, crystals or chunks

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

composition

Dye content, ~97%

composition

-

composition

-

composition

-

assay

≥96.5% (HPLC)

assay

98%

assay

98%

assay

97.5% (GC)

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters