Skip to Content
Merck
All Photos(1)

Key Documents

37419

Supelco

AQUANAL-plus chlorine (Cl2) free and bound 0.01-0.30 mg/L

test set with color comparator and check solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41113319

usage

sufficient for 300 tests

measuring range

0.01-0.30 mg/L (color chart)
0.1-5 mg/L (photometric, Spectro 3)

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Analysis Note

Method
Formation of a red-violet dye N,N-diethyl-1,4-phenylenediamine

Gradation (color chart)
0 - 0.1 - 0.025 - 0.045 - 0.06 - 0.08 - 0.10 - 0.15 - 0.20 - 0.30 mg/L (Cl2)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service