Skip to Content
Merck
All Photos(1)

Key Documents

37194

Supelco

Water hardness (total) test sticks Aquadur®, 3-21°d

Graduation: 3 − 4 − 7 − 14 − 21 °d

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

specific analyte(s)

calcium, magnesium

application(s)

environmental
food and beverages
general analytical

detection method

colorimetric

General description

The hardness of water is expressed in mmol/l or in the older common unit degrees of hardness, whereby it is important to differentiate between German, English and French interpretation:

in Germany: 1 °d = 10 mg/l CaO = 17.8 mg/l CaCO3
in Great Britain: 1 °e = 14.3 mg/l CaCO3
in France 1 °f = 10 mg/l CaCO3

Conversion factors: 1 °d = 1.25 °e = 1.78 °f = 0.178 mmol/l
In the USA, the water hardness is simply expressed in terms of mg/l CaCO3. Apart from the above values, water is often simply classified as "soft water", or "hard water" etc. The following values generally apply to these terms:

below 90 mg/l CaCO3 - very soft water
90 − 180 mg/l CaCO3 - soft water
180 - 270 mg/l CaCO3 - medium hard water
270 - 450 mg/l CaCO3 - hard water
above 450 mg/l CaCO3 - very hard water

The hardness of water is often of importance where no laboratory facilities for the analytical determination exist. In these cases Aquadur® provides a quick method for an indicative reading. Its sensitivity is adequate for textile establishments of moderate size, in dyeing, laundries, small water works, boiler rooms as well as in pisciculture, aquaria and also for household use.

Packaging

test sticks, package with 100 pieces

Legal Information

Aquadur is a registered trademark of Macherey-Nagel GmbH & Co., KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service