Skip to Content
Merck
All Photos(1)

Documents

36564

Supelco

Crimidine

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-4-dimethylamino-6-methylpyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10ClN3
CAS Number:
Molecular Weight:
171.63
Beilstein:
127995
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)c1cc(C)nc(Cl)n1

InChI

1S/C7H10ClN3/c1-5-4-6(11(2)3)10-7(8)9-5/h4H,1-3H3

InChI key

HJIUPFPIEBPYIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crimidine is a convulsant drug, which is widely used as a rodenticide.

Application

Crimidine may be used as an analytical reference standard for the quantification of the analyte in biological samples using different analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J T Lumeij et al.
The Veterinary quarterly, 5(3), 107-111 (1983-07-01)
The diagnosis and treatment of a case of crimidine poisoning in a dog are described. Presenting signs were seizures and vomiting. The vomitus contained a purple coloured cereal grain. The signs and grain colour were suggestive for a rodenticide with
P De Saqui-Sannes et al.
Journal of analytical toxicology, 20(3), 185-188 (1996-05-01)
In an attempt to improve the sensitivity and selectivity of thin-layer chromatographic analysis for the detection of strychnine and crimidine in biological samples, a rapid high-performance thin-layer chromatographic (HPTLC) method with densitometry is described. Fortified dog serum and stomach content
T Besnard et al.
Acta clinica Belgica, 57 Suppl 1, 8-11 (2002-04-27)
Crimidine (2 chloro, 4 methyl, 6 dimethyl amidopyrine) is a synthetic rodenticide which causes acute poisonings after oral ingestion in human. Major toxic effects are consciousness disorders, hypertonic coma and convulsions. Toxic level in human is about 5 mg/Kg. An
[Single dose of 2 acute rodenticides used in a village].
Z Q Sun
Zhonghua liu xing bing xue za zhi = Zhonghua liuxingbingxue zazhi, 6(5), 296-299 (1985-10-01)
A screening procedure for the determination of 13 oral anticoagulants and rodenticides
Lotfi H, et al.
Journal of Analytical Toxicology, 20(2), 93-100 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service