Skip to Content
Merck

35807

4,5-Dichloro-3H-1,2-dithiol-3-one

technical, ≥90% (AT)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3Cl2OS2
CAS Number:
Molecular Weight:
187.07
EC Number:
214-754-5
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
1342220
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

technical

assay

≥90% (AT)

mp

52-56 °C

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C(=O)SS1

InChI

1S/C3Cl2OS2/c4-1-2(5)7-8-3(1)6

InChI key

QGSRKGWCQSATCL-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library