Skip to Content
Merck

Skip To

35382

Secnidazole hemihydrate

VETRANAL®, analytical standard

Synonym(s):

α,2-Dimethyl-5-nitro-1H-imidazole-1-ethanol hemihydrate, (RS)-1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol hemihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
CZK 6,250.00

About This Item

Empirical Formula (Hill Notation):
C7H11N3O3 · 0.5H2O
CAS Number:
Molecular Weight:
194.19
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
8791511

CZK 6,250.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O.CC(O)Cn1c(C)ncc1[N+]([O-])=O.CC(O)Cn2c(C)ncc2[N+]([O-])=O

InChI

1S/2C7H11N3O3.H2O/c2*1-5(11)4-9-6(2)8-3-7(9)10(12)13;/h2*3,5,11H,4H2,1-2H3;1H2

InChI key

LPCFEXQLJSCCMQ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
353873254234196
grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

format

neat

format

neat

format

neat

format

neat

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®

product line

-

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

clinical testing

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters