Skip to Content
Merck
All Photos(1)

Key Documents

34551

Supelco

Dichlorprop-isooctyl

PESTANAL®, analytical standard

Synonym(s):

2,4-DP isooctyl ester, Dichlorprop-isooctyl ester, Isooctyl 2-(2,4-dichlorophenoxy)propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24Cl2O3
CAS Number:
Molecular Weight:
347.28
EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

format

neat

InChI

1S/C17H24Cl2O3/c1-4-5-6-7-8-12(2)21-17(20)13(3)22-16-10-9-14(18)11-15(16)19/h9-13H,4-8H2,1-3H3

InChI key

RQRUQMZZBOQUNB-UHFFFAOYSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service